Learn More
High Five™ SMS Coveralls
Catalog No. 19156222 Shop All Ansell Products
Change view
Click to view available options
Material:
SMS Fabric
SMS Polypropylene
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size
19-156-222 SMS Polypropylene 2X-Large
19-156-220 SMS Polypropylene Large
19-156-221 SMS Polypropylene X-Large
19-156-223 SMS Polypropylene 3X-Large
19-156-224 SMS Polypropylene 4X-Large
19-167-041 SMS Fabric Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-156-222 Supplier High Five™ Supplier No. EW151032XL
This item is not returnable. View return policy
Durable three-layer construction
  • Heavy-duty coveralls
  • Provide protection from many dry particulates and water-based liquids
  • Roomy design
  • No pockets
  • Elastic wrists and ankles
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material SMS Polypropylene
Color White
Disposable Single-use
Cuff Style Elastic
Closure Type Front Zipper
Sterility Non-sterile