Description
- Heavy-duty coveralls
- Provide protection from many dry particulates and water-based liquids
- Roomy design
- No pockets
- Elastic wrists and ankles
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | 3X-Large |
| Gender | Unisex |
| Material | SMS Polypropylene |
| Color | White |
| Disposable | Single-use |
| Cuff Style | Elastic |
| Closure Type | Front Zipper |
| Sterility | Non-sterile |