Learn More
High Five™ SMS Coveralls
Catalog No. 19167041 Shop All Ansell Products
Change view
Click to view available options
Material:
SMS Fabric
SMS Polypropylene
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size
19-167-041 SMS Fabric Medium
19-156-222 SMS Polypropylene 2X-Large
19-156-220 SMS Polypropylene Large
19-156-221 SMS Polypropylene X-Large
19-156-223 SMS Polypropylene 3X-Large
19-156-224 SMS Polypropylene 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-167-041 Supplier High Five™ Supplier No. EW15103M
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Durable three-layer construction
  • Heavy-duty coveralls
  • Provide protection from many dry particulates and water-based liquids
  • Roomy design
  • No pockets
  • Elastic wrists and ankles
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material SMS Fabric
Disposable Single-use
Cuff Style Elastic
Sterility Non-sterile