Learn More
Description
Breathable for all day comfort
- Protect against dry particulates and water-based liquids
- Knit collar and cuffs
- Five-snap front closure
- Available in white or blue
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | 3X-Large |
| Gender | Unisex |
| Material | SMS |
| Color | White |
| Closure Type | Snaps |
| No. of Pockets | 3 |
| For Use With (Application) | Protects from dust, dirt, grime, and water based liquids |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.