Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ Spunbond Isolation Gowns

Catalog No. 19168666 Shop All Ansell Products
Change view
Click to view available options
Color:
Yellow
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-168-666 Yellow Large
19-167-917 Yellow X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-168-666 Supplier High Five™ Supplier No. EY10214L

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Durable protection from common dirt, dust and grime, minor splashes and particulates

Spunbond polypropylene material protects arms, chest, shoulders, waist and upper legs from common workplace particulates.

  • Lightweight, breathable fabric keeps workers cool and comfortable
  • Elastic wrists with easy neck and waist ties for wrap-around styling

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protection from common dirt, dust and grime, minor splashes and particulates
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Size Large
Material Spunbond Polypropylene
Color Yellow
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Model AG163
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.