Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ Spunbond Polypropylene Lab Coats

Catalog No. 19167933 Shop All Ansell Products
Change view
Click to view available options
Color:
Navy
White
Size:
2X-Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-167-933 White 2X-Large
19-167-034 White 3X-Large
19-167-035 White 4X-Large
19-156-183 Navy 2X-Large
19-156-184 Navy 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-167-933 Supplier High Five™ Supplier No. EW102042XL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Economical protection from dust, dirt and grime

  • Breathable construction keeps workers cooler
  • No pockets to catch contaminants
  • Elastic wrists
  • Four-snap closure
  • White

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
For Use With (Application) Protection from dust, dirt, and grime
Construction Breathable Construction Keeps Workers Cooler
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.