Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ Spunbond Shoe Covers

Catalog No. 19156237 Shop All Ansell Products
Change view
Click to view available options
Size: Jumbo; 17 x 7 in.; 300/case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-156-237 Size: Jumbo; 17 x 7 in.; 300/case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-156-237 Supplier High Five™ Supplier No. AS822
This item is not returnable. View return policy

Specifications

Material Polypropylene
Latex Free Yes
Color White
Closure Type Elastic Ankle
For Use With (Application) Ideal to provide protection against general splash and spills
Product Type Shoe Covers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.