Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ High Performance Green Masking Tape

Catalog No. 19072049 Shop All 3M Products
Change view
Click to view available options
Quantity:
16 Rolls/Cs.
48 Rolls/Cs.
Dimensions (L x W):
55 m x 18 mm
55 x 36 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dimensions (L x W)
19-072-049 16 Rolls/Cs. 55 x 36 mm
19-072-047 48 Rolls/Cs. 55 m x 18 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-049 Supplier 3M™ Supplier No. 7000124897
  • $433.00
    $288.65 / Case of 19

    Save $144.35

    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ High Performance Green Masking Tape is ideal for critical paint masking applications in automotive, specialty vehicle, and industrial markets where strong holding and high temp performance is required.

  • Removes cleanly in one piece with no adhesive residue
  • Remains in place around corners, over contours and irregular surfaces
  • Resists paint bleed-through to help create sharp paint lines
  • Green backing improves visibility and accuracy during paint masking
  • Withstands surface temperatures up to 250 degrees F/ 121 degrees C for 30 minutes in paint bake operations
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Rubber
Product Type Masking Tape
For Use With (Application) Masking for Painting, Paint Preparation Masking, Painting and Depainting
Model 401+
Quantity 16 Rolls/Cs.
Thickness (English) 6.7 mil
Dimensions (L x W) 55 x 36 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.