Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ High Performance Masking Tape 232

Catalog No. 19072050 Shop All 3M Products
Change view
Click to view available options
Quantity:
48 Rolls/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-050 48 Rolls/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-050 Supplier 3M™ Supplier No. 7000048942
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ High Performance Masking Tape 232 is capable of masking paint while removing cleanly. Its thick crepe paper backing contains a solvent-resistant saturant that helps resist paint bleed against latex- and oil-based paints.

  • Sticks immediately and resists lifting or curling for paint applications
  • Conforms and maintains integrity when formed around a corner
  • Crepe backing is easy to tear, resists slivering, and provides one piece removal
  • Manufactured without solvents for minimal environmental impact
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Material Rubber
Product Type Industrial Tape
For Use With (Application) Automotive, Design and Construction, Energy, Manufacturing, Mining, Oil and Gas, Transportation
Model 232
Quantity 48 Rolls/Cs.
Thickness (English) 6.3 mil
Dimensions (L x W) 55 m x 18 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.