Learn More
Restek™ High-Power Electronic Crimping Tool with OLED Display Screen
SDP
Catalog No. 06611687 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Power Supply:
Power Cord (China)
Power Cord (Europe)
Power Cord (Japan)
Power Cord (Taiwan)
Power Cord (United Kingdom)
Power Cord (United States)
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Power Supply
06-611-687 Power Cord (Europe)
06-611-684 Power Cord (United Kingdom)
06-611-683 Power Cord (United States)
06-611-688 Power Cord (China)
06-611-686 Power Cord (Japan)
06-611-685 Power Cord (Taiwan)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-611-687 Supplier Restek™ Supplier No. 23236EU
Fixed power supply provides steady, reliable power; Cords are available for China, Europe, Japan, Taiwan, United Kingdom, and United States.
  • Buy only the size crimper or decapper needed (11 mm or 20 mm) with interchangable jaw sets, sold separately
  • The separate crimper and decapper jaw sets fit standard lab vial seals that are aluminum or two-part aluminum/steel caps
  • Ergonomic design reduces strain and arm injury associated with many manual crimping tools
  • Optimize crimping power for each cap type, monitor battery use, and view lifetime statistics with the crisp OLED screen display
  • Save time and minimize adjustments by storing separate programs for different caps and seals
  • Strong enough for use with all steel and magnetic caps
  • Combine with optional base and mounting kit for hands-free operation
  • Two convenient sleep mode options
Specifications
Includes Universal Fixed Power Supply, USB Drive
No. per Pack 1 Ea.
Power Supply Power Cord (Europe)
Type Crimping Tool
Size 11 or 20 mm
For Use With (Application) Crimp Caps