Learn More
Climet Instruments Climet Instruments CI-302 High Pressure Diffuser, Stainless Steel Construction, Compatible With High-Pressure Gases, 1 Each
Supplier: Climet Instruments CI302
The CLIMET INSTRUMENTS CI-302 High Pressure Diffuser is designed for safe and accurate sampling of high-pressure gases. It prevents sensor turbulence and false counts, ensuring reliable performance. Made from stainless steel with no moving parts, it is durable and easy to maintain. This diffuser operates across multiple gas types and supports standard decontamination methods, making it ideal for laboratory and cleanroom environments.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.