Learn More
Norell Inc 5mm, 500MHz 7" EXTREME SERIES LEVEL 3 VALVED THIN WALL NMR TUBE FOR HIGH PRESSURE, 1 TUBE PER PACK
SDP
Supplier:  Norell Inc S5500EX3HPV7
Encompass_Preferred
Level 1 includes an additional fluoroelastomer o-ring seal to augment the existing fluoroelastomer o-ring, thus providing a secondary, backup o-ring if the primary one leaks or fails because of wear or physical damage. Level 3 incorporate additional chemical and solvent resistance over Level 1 by the substitution of Kalrez perfluoroelastomer o-ring(s) in place of the fluoroelastomer o-rings. Kalrez perfluoroelastomer is made from tetrafluoroethylene, and is virtually chemically identical to PTFE fluoropolymer, but physically Kalrez behaves as an elastomeric material, unlike PTFE. Kalrez furnishes the same unsurpassed chemical resistance and degree of inertness as that of PTFE, while affording a higher service temperature limit (up to 288°C compared to 260°C for PTFE) and no propensity to cold flow (or distort under a constant mechanical force or pressure).
Catalog No. NC2832793
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.