Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva High Purity rNTP Set

Catalog No. 45001338 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4 x 25 μmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-338 4 x 25 μmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-338 Supplier Cytiva Supplier No. 27202501
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use for in vitro transcription or for reactions requiring rNTPs of high purity. Cytiva Life Sciences™ High Purity rNTP Set is easy-to-use and offers time-saving solutions.

  • Each lot is tested for ribonuclease contamination to ensure success in your application
  • High purity: >98% triphosphate
  • In a convenient set containing ATP, CTP, GTP, and UTP in separate tubes

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Description Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
pH 7.5
Product Type rNTP Set, 100 mM Solutions (ATP, CTP, GTP, UTP)
Concentration 100 mM
Purity ≥98% triphosphate
Quantity 4 x 25 μmol
For Use With (Application) in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.