Learn More
INTRAVENOUS QUALITY ASSURANCE (IVQA) Category 3 CSP Media-Fill Test Kit
Catalog No. NC1278175
Encompass_Preferred
The Category 3 CSP Media-Fill Test Kit validates compounding personnel's aseptic technique and sterile compounding proficiency. Intended for simulating Category 3 compounded sterile preparations, it evaluates the risk of microbial contamination and assesses competency in aseptic techniques. Suitable for the initial assessment and every 3-month revalidation of aseptic manipulation competency, in accordance with USP Chapter <797> guidelines.Kit Contains:•9 - Sterile 10ml Empty Serum Vials•1 - Container of Tryptic Soy Broth Non-Sterile Powder•1 - Whirl-Pak Bag•1 - Log Sheet
Catalog No. NC1278175 Supplier INTRAVENOUS QUALITY ASSURANCE (IVQA) Supplier No. MFT03
The Category 3 CSP Media-Fill Test Kit validates compounding personnel's aseptic technique and sterile compounding proficiency. Intended for simulating Category 3 compounded sterile preparations, it evaluates the risk of microbial contamination and assesses competency in aseptic techniques. Suitable for the initial assessment and every 3-month revalidation of aseptic manipulation competency, in accordance with USP Chapter <797> guidelines.Kit Contains:•9 - Sterile 10ml Empty Serum Vials•1 - Container of Tryptic Soy Broth Non-Sterile Powder•1 - Whirl-Pak Bag•1 - Log Sheet