Learn More
3M™ Petrifilm™ High-Sensitivity Coliform Count Plate, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S30722 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
50 Pack
500 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30722 50 Pack
S30689 500 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30722 Supplier 3M™ Petrifilm™ Supplier No. 6405
Add to Cart
Add to Cart
Accommodates 5 mL samples and detection of low levels of confirmed coliforms in 24 hours
  • Coliform results in 24 hours
  • Provide accurate, efficient, consistent results for food processors and contract labs
  • Plates are sample-ready, eliminating time-consuming steps of preparing media/agar dishes
  • Unique plate design to trap gas produce by coliform
  • This plate has been awarded Official Methods of Analysis (OMA) #996.02 for Dairy products and NF VALIDATION certificate: 3M-01/7-03/99 for Human food products by AFNOR Certification
Specifications
Detects Coliform
Product Type Count Plate
Test Type Environmental Test
For Use With (Application) Food processors and contract labs, 5 mL coliform samples
Quantity 50 Pack