Learn More
3M™ Petrifilm™ High-Sensitivity Plate Spreader 6481, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S30740 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
2 Pk.
Catalog No. Quantity
S30740 2 Pk.
1 options
Catalog No. S30740 Supplier 3M™ Petrifilm™ Supplier No. 6481
Add to Cart
Add to Cart
Distributes sample evenly for 3M™ Petrifilm™ High-Sensitivity Coliform Count Plates
  • Clear acrylic material allows technician to observe and control spread
  • Spreads inoculum over uniform area
Specifications
Product Type Plate Spreader
For Use With (Equipment) 3M™ Petrifilm™ High-Sensitivity Coliform Count Plates
Quantity 2 Pk.