Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBF Bioscience High-Throughput Submarine Horizontal Electrophoresis Systems
SCI_ED

Catalog No. S22908 Shop All CBS Scientific Products
Change view
Click to view available options
Gel Dimensions:
14 x 26 cm
40 x 26 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Dimensions
S22908 40 x 26 cm
S22906 14 x 26 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S22908 Supplier MBF Bioscience Supplier No. HSU2640

missing translation for 'validMainframeMsg'

Run up to 816 samples with these units.

CBS Economy Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.

Gel Tray Specifications:

  • Winged gel trays for easy handling - no more sticking fingers into slippery buffer to extract gel tray
  • Made from sturdy 6.4mm thick UV transparent acrylic
  • Grooves in gel tray corners create agarose anchors to prevent gel flotation during electrophoresis
  • Gel tray configuration allows single run or multiple simultaneous short runs
  • Three options for gel casting; tapered baffles, external gel casters or continuously adjustable gel caster
  • All combs are adjustable height and have an aluminum backing to keep the combs perfectly straight during gel casting with hot agarose
  • All combs come with microtitre format to allow multi-channel pipetting for accuracy and speed
  • Comb profiles are diagrammed at right to show many configurations, with or without markers
  • PTFE-coated combs allow faster well formation and are recommended when using low percentage or low melt agarose
  • Buffer recirculation ports allow maintenance of uniform pH and ionic strength and reduce frequency of buffer changes necessary during long runs.
  • Tight-fitting safety cover reduces condensation during electrophoresis
  • Coolant can be recirculated beneath gel bed to provide constant temperature runs
  • Combs are printed with catalog number and thickness for easy identification

Applications:
Rapid Screening fo cloning products, rapid screening of restriction fragments, rapid screening of synthetic oligos, rapid screening of PCR products

Specifications

Certifications/Compliance CE
Color Blue
Description Compatible Gel Tray: WGT-3400
Type Horizontal Gel Electrophoresis System
Gel Dimensions 40 x 26 cm
Volume (Metric) Buffer 3400 mL
Warranty 3 Years
Dimensions (L x W) Gel Tray I.D.: 40 x 24.7 cm (15.7 x 9.7 in.)
Dimensions (L x W x H) 64 x 28 x 10 cm (25.1 x 11 x 3.9 in.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.