Learn More
Traction Plus™ High Traction Floor Cleaner and Degreaser
SDP
Catalog No. 18366490 Shop All New Pig Corporation Products
Change view
Click to view available options
Size:
1 gal.
Catalog No. Size
18-366-490 1 gal.
1 options
Catalog No. 18-366-490 Supplier Traction Plus™ Supplier No. CLN9001
Add to Cart
Add to Cart
Pre-treatment cleaner for degreasing even saturated porous floors to enhance their traction.
  • Traction Plus pre-treatment cleaner improves traction by degreasing slippery floors
  • National Floor Safety Institute-certified
  • Penetrates pores and tile surface to release slippery film
  • Safely treat commercial-grade porcelain tile, ceramic tile, quarry tile, terrazzo, concrete and other low-gloss floors
  • Less harsh than most heavy-duty degreasers
  • Great for food-industry floors with heavy grease or for pre-treating floors before applying adhesive-backed mats
  • Strengthens tile surface and grout joints
  • Concentrated to give you more applications
  • Recommended dilutions: 64:1 for light grease, 32:1 for medium grease and 16:1 for heavy grease
  • Do not use on marble, may dull high-gloss floors
  • 1 gal. container
Specifications
Product Type Cleaner/Degreaser
Color-coded No
Form Liquid
Scent Regular Scent
Sterility Non-sterile
Disposable Limited-use
Size 1 gal.
Volume (English) 1 gal.