Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company High Vacuum In Line Filter
SCI_ED

Catalog No. S2337149 Shop All Fischer Technical Company Products
Change view
Click to view available options
Flow Rate:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
S2337149 None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S2337149 Supplier Fischer Technical Company Supplier No. LAVINLINEFILTER
Add to Cart
Add to Cart
Prop 65

In Line Air Filter, features 24 square inches of filter media strong up to 10 CFM air flow.

  • The Inline Air Filter features 24 square inches of filter media which remains strong up to 10cfm air flow.
  • Removes particles as small as 5 microns.
  • For use with 0.5 in. diameter vacuum tubing

Certifications

SciEd

Specifications

Description 24 square inches of filter media strong
Flow Rate None
Pump Type High Vacuum Pump
Applications High Vacuum
Maximum Pressure None
For Use With (Equipment) 0.5 in. Diameter Vacuum Tubing
Voltage 0/1 V
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.