Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries V10AM0322ZL5X
Encompass_Preferred
Class 2 - FR/AR Static Dissipative Mesh Hi-Vis Vest with Orange Modacrylic Binding \nBuilt with GlenGuard? Hi-Vis Static Dissipative Modacrylic fabric which is patented and inherently flame resistant. Designed to make safety vests cooler and more comfortable without sacrificing protection. The fabric is designed with a 10mm carbon core grid-shaped fiber which works to dissipate a potentially harmful static charge allowing safe work in potentially combustible environments.\n\nThe 2" 3M Scotchlite Retroreflectivity Material helps the eye perceive objects in low-light conditions when illuminated by a light source.\n \n ANSI 107-2015 Class 2 \n 2" 3M Scotchlite Reflective Material \n Non-conductive zipper front closure \n Open sides with adjustable snap closure \n 27" length \n\nLakeland'style number? V-10AM0322ZL features static Dissipative properties in a FR/ARC rated mesh modacrylic fabric.
Catalog No. 50-284-1047