Learn More
Lakeland Industries Nomex Coverall
Supplier: Lakeland Industries C010205430
Lakeland Inherently FR 4.5 oz. Nomex coveralls are built tough and comfortable to protect the wearers from flash fires. C010 coveralls are configured with a quick release zipper, heavy duty snaps, pockets, and an elasticized waistband. Available in Navy and Khaki, sizes SM-6X, this coverall is ideal when flash fire hazards are present.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.