Learn More
New Pig High-Visibility Absorbent Mat Pad
SDP
Catalog No. 17100912 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W):
20 x 15 in.
Catalog No. Dimensions (L x W)
17-100-912 20 x 15 in.
1 options
Catalog No. 17-100-912 Supplier New Pig Supplier No. MAT604
Add to Cart
Add to Cart
Hard-to-miss high-visibility yellow mat draws attention to spill cleanups in progress and alerts workers of potential hazards.
  • Hard-to-miss high-visibility yellow mat draws attention to spill cleanups in progress and alerts workers of potential hazards.
  • Lightweight mat pads are for minor messes, small leaks and drips or lining tool carts; dispenser box fits neatly on shelves or carts
  • Bright yellow color stands out, so it's easy for workers to see hazards, repairs, or cleanups in progress, even in poorly lit areas.
  • PIG Mat lasts 2x longer than competitive mats and won't rip, tear or fray even when saturated; liquids absorb quickly for faster, easier cleanups and safer workplaces
  • Absorbs most common industrial liquids - oils, coolants, solvents, water and more
  • Can be wrung out and incinerated after use to reduce waste or for fuel blending.
Specifications
Color Yellow
Material Polypropylene
Type Universal Absorption Pad
Disposable Single-use
Packaging Quantity 200 Pads/Bag
Weight Capacity Lightweight
Dimensions (L x W) 20 x 15 in.