Specifications
| Lens Coating | Anti-fog |
| Lens Type | H2X |
| Certifications/Compliance | ANSI Z87.1, CSA Z94.3-07 |
| Lens Tint | Amber |
| Product Type | Ballistic Safety Glasses |
| Frame Color | Black |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |