Specifications
| Lens Type | H2X |
| Certifications/Compliance | ANSI Z87.1 |
| Certifications/Compliance (Europe) | EN-166 |
| Lens Tint | Gray |
| Frame Color | Black |
| Weight (Metric) | 35 g |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |