Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ HILED Lighting (GM) Upgrade
GREENER_CHOICESDP

Catalog No. 01257247 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Color:
Far Red
Red, Green, Blue and UV
UV
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-257-247 UV
01-257-241 Red, Green, Blue and UV
01-257-246 Far Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-257-247 Supplier Syngene™ Supplier No. GMHILEDUPGDUV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HILED lighting upgrade from RGB or RIR, designed to allow for the addition of UV, Far Red, Red, Green, or Blue.

  • Required equipment: GM-HILED-RGB or GM-HILED-RIR
  • Bag of parts to be fitted by professional
  • Works with GBOX mini only

Specifications

Product Type Lighting
Color UV
For Use With (Equipment) GM-HILED-RGB or GM-HILED-RIR
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.