Learn More
Syngene™ HILED RGB Lighting Upgrade
GREENER_CHOICESDP
Catalog No. 01257244 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
GM-HILED-RGB
Catalog No. For Use With (Equipment)
01-257-244 GM-HILED-RGB
1 options
Catalog No. 01-257-244 Supplier Syngene™ Supplier No. GMHILEDUPGDIR
Add to Cart
Add to Cart
HILED lighting upgrade from RGB to RGBIR.
  • Required equipment: GM-HILED-RGB
  • Bag of parts to be fitted by professional
  • Works with GBOX mini only
Specifications
Product Type Lighting
For Use With (Equipment) GM-HILED-RGB