Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ HILED Upgrade Lighting from RGB to RGBIR
GREENER_CHOICESDP

Catalog No. 01257281 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Color:
Far Red
Infra Red
Red, Green, Blue, UV
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-257-281 Red, Green, Blue, UV
01-257-285 Far Red
01-257-283 Infra Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-257-281 Supplier Syngene™ Supplier No. HILEDRGBUV
Add to Cart
Add to Cart
Request a Quote

Syngene™ HILED Upgrade Lighting from RGB to RGBIR works with the latest GBOX ONLY (not previous modular system).

For use with GBOX

Specifications

For Use With (Application) Multiplex Western Fluorescence
Quantity 1
Color Red, Green, Blue, UV
For Use With (Equipment) Gbox Chemi Systems
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.