Learn More
Genesee Scientific Corporation HINGED CRYOBOX PURPLE 30PK
SDP
Supplier:  Genesee Scientific Corporation 90507P
Encompass
BioCision BCS-219P, Hinged CryoBox, Purple, 30 pack 81 Cryogenic Vials, 30 Cryoboxes/Unit
Catalog No. NC2404156
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.