Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiPrep™ IEX FF 16/10 Columns

Catalog No. 45002070 Shop All Cytiva Products
Change view
Click to view available options
Ligand Type:
Carboxymethyl
Diethylaminoethyl
Max. Pressure:
1.5 bar
5 bar
pH Range:
2 to 12
6 to 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Ligand Type Max. Pressure pH Range
45-002-070 Carboxymethyl 1.5 bar 6 to 10
45-002-066 Diethylaminoethyl 5 bar 2 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-070 Supplier Cytiva Supplier No. 28936542
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provide fast, reliable and reproducible preparative ion exchange separations

DEAE and CM Sepharose™ 6 Fast Flow are prepacked in 20mL HiPrep columns. Ideal for rapid enrichments during initial capture of proteins from starting material.

Features:

  • Combine good flowrates and high binding capacity
  • Simple-to-operate
  • Compatible with single-pump configurations and other chromatography systems (e.g., AKTA™)

Specifications (H2O at room temp.)

  • Dimensions: 1.6 x 10cm
  • Recommended flowrate: 2-10mL/min. (60-300cm/h)
  • Maximum flowrate: 10mL/min. (300cm/hr.)
  • Max. pressure over packed bed: 1.5bar (22psi, 0.15MPa)
  • Column hardware pressure limit: 5bar (73psi, 0.5MPa)
  • Storage: 20% ethanol (DEAE,CM), 0.2M sodium acetate in 20% ethanol (SP)
  • Storage temperature: 4° to 30°C

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
Product Type HiPrep CM Sepharose FF column
Bed Volume 20mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 20mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate >300 cm/hr.
Material Polypropylene
Ligand Type Carboxymethyl
Max. Pressure 1.5 bar
pH Range 6 to 10
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Preparative separations of Proteins and other biomolecules using ion exchange (IEX) chromato graphy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.