Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiPrep™ IMAC FF 16/10 Column

Catalog No. 45002072 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-072 1 x 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-072 Supplier Cytiva Supplier No. 28936552
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For optimizing purification of histidine-tagged proteins at laboratory scale

IMAC Sepharose™ 6 Fast Flow is prepacked in 20mL HiPrep columns for greater scale up. The medium consists of 90μm beads of highly crosslinked agarose, to which a chelating group has been covalently coupled. This group may be charged with suitable metal ions, specific for your target protein.The bead size enables high flow properties; low metal ion leakage retains activity of the purified protein.

Specifications:
IMAC Sepharose 6 Fast Flow

  • Matrix: highly crosslinked agarose, 6%
  • Average particle size: 90μm
  • Binding capacity (protein/ion dependent): approx. 40mg histidine-tagged protein/mL medium when charged with Ni2+
  • Compatibility: stable in all commonly used buffers
  • Chemical stability (uncharged medium) one week, 40°C: 0.01 M HCl, 0.1 M NaOH; 12 hr.: 1M NaOH, 70% acetic acid; 1 hr. tested 2% SDS; 30 min. tested 30% 2-propanol
  • Stability (pH, w/ uncharged medium w/ metal ion): 3 to 12 (long-term), 2 to 14 (short-term)
  • Storage/Temperature: 20% ethanol; 4° to 30°C

HiPrep Prepacked Columns

  • Volume: 20mL
  • Dimensions: 1.6 x 10cm
  • Max. pressure during operation: 1.5bar (0.15MPa, 22psi)
  • Hardware pressure limit: 5bar (0.5MPa, 73psi)
  • Recommend flowrate (H2O at room temp.): 2 to 10mL/min.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Buffer 0.01M HCl, 1M NaOH, 70% Acetic Acid, 2% SDS, 30% 2-propanol
    Product Type HiTrap IMAC Sepharose 6 FF column
    Bed Volume 20mL
    Diameter (English) Inner 0.62 in.
    Diameter (Metric) Inner 16mm
    Quantity 1 x 20mL
    Height (English) 3.93 in.
    Height (Metric) 100mm
    Flow Rate <10mL/min.
    Material Polypropylene
    Max. Pressure 1.5 bar
    pH Range 3 to 12
    Particle Size 45 to 165μm
    Support Type 6% Cross-linked Agarose
    Content And Storage 4° to 30°C
    Column Type Ion Exchange Column
    For Use With (Application) IMAC and subsequent purification of Polyhistidine tagged Proteins
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.