Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiPrep™ Sephacryl™ S-100 HR Columns

Catalog No. 45000206 Shop All Cytiva Products
Change view
Click to view available options
Flow Rate:
<0.5 mL/min.
<1.3 mL/min.
Bed Volume:
120 mL
320 mL
Diameter (Metric) Inner:
1.6cm
2.6cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Bed Volume Diameter (Metric) Inner
45-000-206 <1.3 mL/min. 320 mL 2.6cm
45-000-195 <0.5 mL/min. 120 mL 1.6cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-206 Supplier Cytiva Supplier No. 17119401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maximum recovery and minimum nonspecific adsorption

  • Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Recommended linear flowrate 5 to 15cm/hr.

Designed for separating peptides and small proteins

TRUSTED_SUSTAINABILITY

Specifications

Quantity Each
Flow Rate <1.3 mL/min.
Particle Size 25 to 75 μm
For Use With (Equipment) Low to medium pressure AKTA™ systems
Bed Volume 320 mL
Product Type Sephacryl HR Column
Diameter (Metric) Inner 2.6cm
For Use With (Application) Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix
Height (English) 23.62 in.
Height (Metric) 60cm
Material Polypropylene
Max. Pressure 1.5 bar [0.15 MPa] (22 psi)
pH Range 3 to 11
Support Type Allyl Dextran and N, N'-methylenebisacrylamide
Sample Type Peptides and small proteins
Content And Storage 4°C to 30°C, 20% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.