Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiPrep™ Sephacryl™ S-300 HR Columns

Catalog No. 45000197 Shop All Cytiva Products
Change view
Click to view available options
Flow Rate:
<0.5 mL/min.
<1.3 mL/min.
Bed Volume:
120mL
320mL
Diameter (Metric) Inner:
1.6cm
2.6cm
Format:
Bed Dimensions: 16x60
Bed Dimensions: 26x60
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Bed Volume Diameter (Metric) Inner Format
45-000-197 <0.5 mL/min. 120mL 1.6cm Bed Dimensions: 16x60
45-000-208 <1.3 mL/min. 320mL 2.6cm Bed Dimensions: 26x60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-197 Supplier Cytiva Supplier No. 17116701
Add to Cart
Add to Cart

Maximum recovery and minimum nonspecific adsorption

  • Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Recommended linear flowrate 5 to 15cm/hr.
  • Designed for purifying antibodies, serum proteins and midsize proteins

TRUSTED_SUSTAINABILITY

Specifications

Quantity Each
Flow Rate <0.5 mL/min.
Particle Size 25 to 75 μm
For Use With (Equipment) Low to medium pressure AKTA™ systems
Bed Volume 120mL
Product Type Sephacryl HR Column
Format Bed Dimensions: 16x60
Diameter (Metric) Inner 1.6cm
For Use With (Application) Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix
Height (English) 23.62 in.
Height (Metric) 60cm
Material Polypropylene
Max. Pressure 22psi
pH Range 3 to 11
Support Type Allyl Dextran and N, N∽-methylenebisacrylamide
Sample Type Antibodies, serum proteins and mid-size proteins
Content And Storage 4° to 30°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.