Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiPrep™ SP HP 16/10 Column

Catalog No. 45000556 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-556 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-556 Supplier Cytiva Supplier No. 29018183
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provide fast, preparative separations of proteins and other biomolecules

  • Strong cation exchanger designed for preparative ion exchange chromatography separations
  • Highly cross-linked agarose provides excellent chemical and physical stability
  • Slim, modern design allows for easy handling and connectionl

TRUSTED_SUSTAINABILITY

Specifications

Description HiPrep SP HP 16/10
Diameter (English) Inner Column 0.6 in.
Flow Rate 1 to 5 mL/min.
For Use With (Application) HiPrep SP HP 16/10 is a strong cation exchanger designed for preparative ion exchange chromatography separations. This 20 mL column is prepacked with SP Sepharose High Performance chromatography resin. The resin is based on rigid, highly cross-linked bead
Content And Storage 4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 20 mL
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.