Learn More
Genscript Corporation HIRUDIN RECOMBINANT PROTEIN
Supplier:  Genscript Corporation Z028121
Encompass_Preferred
Hirudin is a potent thrombin inhibitor originally derived from the medicinal leech Unlike heparin, hirudin act directly on thrombin, rather than through other clotting factors. They have a high binding affinity and specificity for thrombin. The mechanism of hirudin-thrombin binding appears to be unique. Recombinant hirudin variant is derived from yeast cell, the polypeptide containing 65 amino acid residues has a molecular weight of 6979.5 Da, which is identical to natural hirudin except for substitution of leucine for isoleucine at the N-terminal end of the molecule and the absence of a sulfate group on the tyrosine at position 63.
Catalog No. 50-276-608
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.