Learn More
Cytiva His Mag Sepharose™ Ni Magnetic Beads
Catalog No. 45002514 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 x 1 mL
Catalog No. Quantity
45-002-514 2 x 1 mL
1 options
Catalog No. 45-002-514 Supplier Cytiva Supplier No. 28967388
Add to Cart
Add to Cart
Designed for efficient, small-scale purification/screening of histidine-tagged proteins from different sources Cytiva Lifescience™ His Mag Sepharose™ Ni Magnetic Beads enable high-capacity small-scale purification.
Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel. For higher throughput, larger numbers of samples can easily be screened in parallel on a robotic device.
  • High-capacity small-scale purification of histidine-tagged proteins from different sources
  • High purity and yield
  • Easy parallel screening of histidine-tagged proteins with high repeatability
  • Scalability dash (en) Simple capture of histidine-tagged proteins from small or large sample volumes (low microliter to milliliter scale)
TRUSTED_SUSTAINABILITY
Specifications
Binding Property 50 mg Histidine-tagged protein/mL medium
Quantity 2 x 1 mL