Learn More
Description
- No filtration needed: load unclarified sample directly, increasing reproducibility and saving time
- Compatible with robotic systems or manual protocols with centrifuge or vacuum
- Easy scale-up
- Long-term pH range 3 to 12
- Protein binding capacity up to 800μg histidine-tagged protein/well with His MultiTrap FF
- Protein binding capacity up to 1mg histidine-tagged protein/well with His MultiTrap HP
Specifications
Specifications
| Description | His MultiTrap FF and His MultiTrap HP are prepacked disposable 96-well plates for reproducible high throughput parallel purification of histidine-tagged recombinant proteins by Immobilized Metal ion Affinity Chromatography (IMAC) |
| Material | Polypropylene, Polyethylene |
| pH Range | 3 to 12 |
| Support Type | 6% Highly Cross-linked Agarose |
| Content And Storage | 4°C to 30°C |
| Particle Size | 34 μm |
| Quantity | 4 x 96-well Filter Plates |
| Type | 96-well Filter Plate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.