Learn More
MONOD BIO INC Anti-Histidine NovoBody Binder Protein
Supplier:  MONOD BIO INC MB7973
Encompass
Our NovoBody™ technology leverages state-of-the-art protein design to amplify the strengths of antibodies while addressing their limitations. NovoBody™ binders meet the evolving demands of research and diagnostics, offering the power to develop assays with unmatched speed, performance, and the flexibility to take your products to the next level. Tag: Twin-Strep-tagConjugation: NoneApplications: ELISA, BLI, CaptureAmount: 100 micrograms
Catalog No. NC3705664
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.