Learn More
MBL International Corporation His tag peptide
SDP
Supplier:  MBL International Corporation 3310205
Encompass_Preferred
Size:2 mg x 5; Target:His; This synthetic peptide including 6xHis tag sequence has been designed for the elution of His tagged Protein PURIFICATION KIT. The peptide sequence is XXX-(6xHis)-XXX.His tag peptide.
Catalog No. 50-446-61
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.