Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Capto™ adhere Multimodal Prepacked Column

Catalog No. 45002532 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 4.7mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-532 1 x 4.7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-002-532 Supplier Cytiva Supplier No. 28926981
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For intermediate purification and polishing of monoclonal antibodies after the protein A capture step

  • Strong multimodal anion exchanger with N-benzyl-n-methyl ethanolamine ligand
  • Offers alternative selectivity compared to traditional anion exchangers
  • In flowthrough mode, removal of leached protein A, aggregates, host cell proteins, nucleic acids and viruses from MAbs is performed
  • Antibodies pass directly through the column while the contaminants remain bound
  • Highly cross-linked agarose matrix
  • Ready-to-use columns for convenient process development optimization
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetic Acid, 1M NaOH
Product Type HiScreen Capto adhere column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <600cm/hr.
Material Polypropylene
Ligand Type N-benzyl-n-methyl Ethanolamine
Max. Pressure 3 bar
pH Range 3 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Multimodal Column
For Use With (Application) Optimization and parameter screening
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.