Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen Capto Blue

Catalog No. 45002551 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
7.7 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
45-002-551 7.7 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-551 Supplier Cytiva Supplier No. 28992474
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Excellent for method optimization and parameter screening due to the 10 cm bed height

  • Prepacked with Capto Blue
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same flow velocity
TRUSTED_SUSTAINABILITY

Specifications

Description HiScreen Capto Blue
Bed Volume 4.7 mL
Diameter (Metric) Inner 7.7 mm
Flow Rate 1.2 to 2.3 mL/min.
Max. Pressure 3 bar [0.3 MPa] (44 psi)
Content And Storage 2° to 8°C
Column Type Affinity Column
For Use With (Application) Group specific affinity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.