Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen Capto™ L Columns
GSA_VA

Catalog No. 45002604 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-604 4.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-604 Supplier Cytiva Supplier No. 17547814
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HiScreen Capto L columns are prepacked with Capto L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.

HiScreen Capto L columns are part of the process development platform available from Cytiva.

  • Prepacked, ready-to-use columns for convenient process development optimization.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume (4.7ml) gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Flow Rate <500 cm/hr.
Material Polypropylene
pH Range 2 to 10
Support Type Rigid, highly cross-linked Agarose
Content And Storage 2°C to 8°C
Particle Size 85 μm
Quantity 4.7 mL
For Use With (Application) Optimization and parameter screening
Type Agarose
Ligand Type Recombinant Protein L (E. coli), mammalian free
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.