Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Capto™ MMC Multimodal Prepacked Column

Catalog No. 45002531 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 4.7mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-531 1 x 4.7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-531 Supplier Cytiva Supplier No. 28926980
Add to Cart
Add to Cart

Allows binding of proteins at the conductivity of the feed material and solves specific purification challenges

  • With innovative salt-tolerant ligand
  • Weak multimodal cation exchanger with a different selectivity compared with traditional ion exchangers
  • Highly cross-linked agarose matrix
  • Ready-to-use columns for convenient process development optimization
  • Bed height of 10cm HiScreen columns is designed to allow method optimization and parameter screening
  • Can be easily connected in series to achieve 20cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetic Acid, 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride and 70% Ethanol
Product Type HiScreen Capto MMC column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <600cm/hr.
Material Polypropylene
Max. Pressure 3 bar
pH Range 2 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Multimodal Column
For Use With (Application) Optimization and parameter screening
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.