Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Capto™ Phenyl (High Sub) Prepacked HIC Column

Catalog No. 45002549 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 4.7mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-549 1 x 4.7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-549 Supplier Cytiva Supplier No. 28992472
Add to Cart
Add to Cart

Hydrophobic interaction chromatography (HIC) media used in the capture and intermediate stages of protein purification, used for method optimization and parameter screening

  • Highly cross-linked agarose matrix with average bead size of 75μm
  • Has good flow properties and high loading capacities
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • Connect easily in series to achieve 20cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Capto Phenyl (high sub) media is used for initial and intermediate step purifications requiring a chromatography medium with medium to high hydrophobicity
  • Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

  • Bed volume: 4.7mL
  • Bed dimension: 7.7 × 100mm
  • Flow rate: 1.2 to 2.7mL/min.
  • Flow velocity: <600cm/hr.
Media
  • Average particle size: 75μm
  • Degree of substitution: 27μmol/mL gel
  • pH stability cleaning: 2 to 14
  • Pressure/Flow specification: 300kPa at 600cm/hr., 1m diameter column, 20cm bed height
  • Medium: BioProcess
  • pH stability working range: 3 to 13
  • Ligand: phenyl
  • Binding capacity/mL chromatography medium: 27mg BSA /mL (at QB10%)
Column
  • Bed support: filter
  • Material (column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type HiScreen Capto Phenyl (high sub) column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <600cm/hr.
Material Polypropylene
Ligand Type Phenyl (∼27μM/mL drained medium)
Max. Pressure 43.5psi
pH Range 3 to 13
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type HIC Column
For Use With (Application) Optimization and parameter screening
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.