Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Capto™ Q ImpRes and Capto™ SP ImpRes

Catalog No. 45002564 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Capto Q ImpRes medium
Capto SP ImpRes medium
Ligand Type:
Quaternary Amine
Sulphonate group
pH Range:
2 to 12
4 to 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Ligand Type pH Range
45-002-564 Capto Q ImpRes medium Quaternary Amine 2 to 12
45-002-562 Capto SP ImpRes medium Sulphonate group 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-564 Supplier Cytiva Supplier No. 17547002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-resolution intermediate purification and polishing based on the well-established Capto platform with traditional ligands

  • Delivers pressure-flow properties with wide resolution
  • High-throughput purifications easy to optimize and scale up
  • High flow agarose based matrix
  • Runs at high flow rates as well as high bed heights thus increasing flexibility in process design
  • Developed for large-scale purification of biopharmaceutical proteins
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
  • Available in a wide range of different bulk pack sizes and pre-packed formats for process development

Capto Q ImpRes: Strong anion (quaternary amine group) exchange BioProcess™ medium
Capto SP ImpRes: Strong cation (sulfonate group) exchange BioProcess medium

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type Capto Q ImpRes medium
Diameter (English) Inner 39.3 in.
Diameter (Metric) Inner 1000mm
Quantity 100 mL
Height (English) 7.87 in.
Height (Metric) 200mm
Flow Rate 220 cm/hr.
Ligand Type Quaternary Amine
Max. Pressure <3 bar
pH Range 2 to 12
Particle Size 36 to 44μm
Support Type High-flow Agarose
Content And Storage 4° to 30°C
For Use With (Application) High-throughput intermediate purification and polishing steps of a wide range of biomolecules
Matrix Capto Q Chromatography Media
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.