Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen Capto Q ImpRes Prepacked Column

Catalog No. 45002524 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
7.7 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
45-002-524 7.7 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-524 Supplier Cytiva Supplier No. 17547015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HiScreen Capto Q ImpRes columns are prepacked with Capto Q ImpRes ion exchange chromatography medium, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

  • Capto Q ImpRes is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 4.7
Diameter (Metric) Inner 7.7 mm
Flow Rate 100 to 300 cm/h
Max. Pressure 4 bar [0.4 MPa] (59 psi)
Content And Storage 4° to 30°C, in 20% Ethanol
Column Type Ion Exchange Column
For Use With (Application) Ion Exchange Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.