Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Capto™ S ImpAct Prepacked Column

Catalog No. 45003204 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 4.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-003-204 1 x 4.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-003-204 Supplier Cytiva Supplier No. 17371747
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HiScreen Capto S ImpAct columns are prepacked with Capto S ImpAct cation exchange chromatography medium, and are part of our process development platform. The columns are an excellent choice for method optimization and parameter screening.

  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Easily connected in series to achieve 20 cm bed height.
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.
  • Prepacked with Capto S ImpAct, a strong cation exchange BioProcess medium, designed for immediate purification and polishing of a wide range of biomolecules, especially monoclonal antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Product Type HiScreen Capto S ImpAct Prepacked Column
Quantity 1 x 4.7 mL
Flow Rate 2.3 mL/min
Particle Size 90μm
Column Type Ion Exchange Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.