Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva™ HiScreen™ Column with MabSelect™ VL Protein L Resin

Catalog No. 01257013 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Column
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-257-013 1 Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-257-013 Supplier Cytiva™ Supplier No. 17542015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These HiScreen™ columns contain Protein L resin for affinity capture of antibody Fabs, bispecifics, and other variants containing a kappa light chain.

As the diversity of antibody variants increases, so does the need for chromatography resins that can capture and purify these variants. As an alternative to protein A, protein L resin can be used to capture antibody Fabs, bispecific antibodies, and other molecules containing kappa variants of the variable light (VL) chain. Protein L binds human kappa light chain subtypes 1, 3, and 4 and mouse kappa light chain subtype 1.

Redesigned protein L ligand MabSelect™ VL protein L resin showed a two-fold increase in binding capacity compared to Cytiva™ Capto™ L chromatography resin. This higher capacity provides productivity advantages in research and biomanufacturing.

  • A redesigned protein L ligand captures antibody Fabs, bispecifics, and other variants containing a kappa light chain with high binding capacity and improved alkaline stability
  • High dynamic binding capacity for efficient capture of target molecules
  • Stable when cleaned with 0.1 M NaOH, reducing risk for bioburden incidents
  • Provides good resolution for product-related impurities in the capture of bispecific antibodies
  • Convenient for method optimization and parameter screening enabled by the 10 cm bed height for efficient chromatography process development
TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 70 mg Human IgG/mL Resin at 6 min. Residence Time, 60 mg Human IgG/mL Resin at 4 min. Residence Time
Capacity (Metric) 4.7 mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7 mm
Flow Rate <300 cm/hr.
Max. Operating Pressure 3 bar
Particle Size 60 μm
Product Line HiScreen MabSelect
Quantity 1 Column
For Use With (Application) Antibody Affinity Chromatography
Target Antibody
Type Pre-packed Column
Format Chromatography Column
Bed Height 100 mm
Column Type Affinity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.