Learn More
Description
This column is prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.
Specifications
Specifications
| Bed Volume | 4.7 mL |
| Flow Rate | <500 cm/hr. |
| Max. Operating Pressure | 3 bar (0.3 MPa, 44 psi) |
| Quantity | 1 x 4.7 mL |
| For Use With (Application) | Antibody affinity chromatography |
| Type | Pre-packed Column |
| Binding Capacity | ≈80 mg Human IgG/mL Resin at 6 min residence time. ≈40 mg Human IgG/mL at 2 min residence time. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.