Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen Phenyl FF (High Sub)

Catalog No. 45002537 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
7.7 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
45-002-537 7.7 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-537 Supplier Cytiva Supplier No. 28926988
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HiScreen Phenyl FF (high sub) columns are prepacked with Phenyl Sepharose 6 Fast Flow hydrophobic interaction chromatography (HIC) media. The columns are an excellent choice for method optimization and parameter screening.

  • Phenyl Sepharose 6 Fast Flow is a well established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.Two levels of substitution help find optimal selectivity and binding capacity for a given application.
  • Prepacked, ready-to-use columns for convenient process development optimization.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity.
TRUSTED_SUSTAINABILITY

Specifications

Description HiScreen Phenyl FF (High Sub)
Bed Volume 4.7
Diameter (Metric) Inner 7.7 mm
Flow Rate Less than 450 cm/h
Max. Pressure 1.5 bar [0.15 MPa] (22 psi)
Content And Storage 4° to 30°C, 20% Ethanol or 0.01M NaOH
Column Type HIC Column
For Use With (Application) Hydrophobic Interaction Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.