Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen Prepacked HIC Column, Capto Butyl

Catalog No. 45002550 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Piece
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-550 1 Piece
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-550 Supplier Cytiva Supplier No. 28992473
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intended for initial and intermediate step purifications that require a medium with low to medium hydrophobicity.

  • Capto Butyl often works efficiently with rather low salt concentrations
  • Small bed volume for fast results and minimal sample/buffer consumption
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Piece
Flow Rate <600cm/h
Material Polypropylene
Ligand Type Butyl
Particle Size 75μm (median particle size of the cumulative volume distribution)
Content And Storage 4° to 30°C, 20% Ethanol
Column Type HIC Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.